Skip to Content
CZ
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Hydrogen ion buffers for biological research.
Hydrogen ion buffers for biological research.
Biochemistry (1966-02-01)
N E Good, G D Winget, W Winter, T N Connolly, S Izawa, R M Singh
PMID
5942950
Product Description
P6757
Sigma-Aldrich
PIPES, ≥99% (titration)
Pricing
69889
Sigma-Aldrich
MES monohydrate, BioUltra, Molecular Biology, ≥99.5% (T)
Pricing
P8203
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Pricing
69890
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Pricing
H0887
Sigma-Aldrich
HEPES solution, 1 M, sterile-filtered, BioReagent
Pricing