Merck
  • Ionic liquid-reconstituted cellulose composites as solid support matrices for biocatalyst immobilization.
Ionic liquid-reconstituted cellulose composites as solid support matrices for biocatalyst immobilization.
Biomacromolecules (2005-09-13)
Megan B Turner, Scott K Spear, John D Holbrey, Daniel T Daly, Robin D Rogers
ABSTRACT
Preparation of cellulose-polyamine composite films and beads, which provide high loading of primary amines on the surface allowing direct one-step bioconjugation of active species, is reported using an ionic liquid (IL) dissolution and regeneration process. Films and bead architectures were prepared and used as immobilization supports for laccase as a model system demonstrating the applicability of this approach. Performance of these materials, compared to commercially available products, has been assessed using millimeter-sized beads of the composites and the lipase-catalyzed transesterification of ethyl butyrate.
Sigma-Aldrich
Syringaldazine, 98%
Sigma-Aldrich
Ethyl butyrate, 99%
Sigma-Aldrich
Syringaldazine, indicator for laccase and peroxidase activity
Sigma-Aldrich
Ethyl butyrate, natural, ≥98%, FCC, FG
Supelco
Ethyl butyrate, analytical standard
Sigma-Aldrich
Ethyl butyrate, ≥98%, FCC, FG