Merck
  • Fluorescence determination of the pesticide asulam by flow injection analysis.
Fluorescence determination of the pesticide asulam by flow injection analysis.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-01-25)
I Súbová, A Khenlami Assandas, M Catalá Icardo, J Martínez Calatayud
ABSTRACT
This paper presents the analytical determination of the pesticide Asulam based on its native fluorescence. The method was optimized in either a flow injection analysis (FIA) assembly or in batch. The maximum fluorescence intensity was observed for basic pH solutions and at a lamda(ex) of 258 nm and a lamda(em) of 342 nm. The influence of different empirical parameters, such as the pH, the presence of surfactants, solvent polarity or solved oxygen amount, was studied. The calibration range was fitted with a linear equation from 0.01-3 mg l(-1) Asulam and 0.005-15 mg l(-1) Asulam for batch and continuous-flow, respectively. The RSD for both procedures was 1.0%. After testing the influence of a large series of potential interferents, the method was applied to water samples from different locations.