- A genotoxicological study of hexachlorobenzene and pentachloroanisole.
A genotoxicological study of hexachlorobenzene and pentachloroanisole.
Teratogenesis, carcinogenesis, and mutagenesis (1991-01-01)
P Siekel, I Chalupa, J Beno, M Blasko, J Novotný, J Burian
PMID1677498
ABSTRACT
The potential mutagenic activity of hexachlorobenzene (HCB) and pentachloroanisole (PCA) was investigated. No genotoxicity after application on Salmonella typhimurium (Ames test), Escherichia coli, and human peripheral blood lymphocytes in vitro was observed.