Merck
  • Allergic contact dermatitis to copolymers in cosmetics--case report and review of the literature.
Allergic contact dermatitis to copolymers in cosmetics--case report and review of the literature.
Contact dermatitis (2006-10-10)
Sarah Quartier, Marjan Garmyn, Sophie Becart, An Goossens
ABSTRACT
Copolymers or heteropolymers are large molecules with high molecular weights (>1000 D). They have been underestimated for a long time as to their sensitizing capacities. Allergic contact dermatitis to 6 copolymers in cosmetics and 1 in a medical dressing has been described; however, the nature of the hapten is still unknown. We report a case of allergic contact dermatitis to polyvinylpyrrolidone (PVP)/hexadecene copolymer in a purple-colored lipstick and review the literature on allergic contact dermatitis to 7 copolymers: PVP/hexadecene, PVP/eicosene, PVP/1-triacontene, methoxy polyethyleneglycol (PEG)-22/dodecyl glycols, methoxy PEG-17/dodecyl glycols, phthalic anhydride/trimellitic anhydride/glycols, and polyvinyl methyl/maleic acid anhydride.
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%