Merck
  • [The production of gastrodin through biotransformation of p-hydroxybenzaldehyde by cell suspension culture of Datura stramonium].
[The production of gastrodin through biotransformation of p-hydroxybenzaldehyde by cell suspension culture of Datura stramonium].
Yao xue xue bao = Acta pharmaceutica Sinica (2006-12-23)
Jia-Shun Gong, Wei-Peng Ma, Jun-Xue Pu, Shu-Guan Xu, Shuang-Qing Zheng, Chun-Jie Xiao
ABSTRACT
To investigate the production of p-hydroxymethylphenol-beta-D-glucoside (gastrodin) through biotransformation by plant cell suspension cultures. Using cell suspension cultures of Datura stramonium to convert the exogenous p-hydroxybenzaldehyde into gastrodin was conducted and the converted compounds were separated with a combination of multi-chromatography. Their chemical structures were determined on the basis of spectral analysis and chemical evidence. The conversion procedure of p-hydroxybenzaldehyde into gastrodin by Datura stramonium cell suspension cultures was established. The synthesized gastrodin (II) was isolated from the fermental liquor and identified by spectral analysis. At the same time, the p-hydroxybenzyl alcohol (I) converted through biotransformation of p-hydroxybenzaldehyde by cell suspension cultures of Datura stramonium was also isolated and identified. Two compounds were also isolated from the cell cultures and they were identified as beta-D-furanoallulose (III) and n-butyloxystyryl-beta-D-pyranoallulose (IV). Datura stramonium grown in suspension cultures can convert exogenous p-hydroxybenzaldehyde into the corresponding gastrodin.
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG
Sigma-Aldrich
4-Hydroxybenzyl alcohol, 99%