Merck
  • Synthesis of some members of the hydroxylated phenanthridone subclass of the Amaryllidaceae alkaloid family.
Synthesis of some members of the hydroxylated phenanthridone subclass of the Amaryllidaceae alkaloid family.
The Journal of organic chemistry (2007-03-07)
Albert Padwa, Hongjun Zhang
ABSTRACT
The total synthesis of several members of the hydroxylated phenanthridone subclass of the Amaryllidaceae alkaloid family has been carried out. (+/-)-Lycoricidine and (+/-)-7-deoxypancratistatin were assembled through a one-pot Stille/intramolecular Diels-Alder cycloaddition cascade to construct the core skeleton. The initially formed [4+2]-cycloadduct undergoes nitrogen-assisted ring opening followed by a deprotonation/reprotonation of the resulting zwitterion to give a rearranged hexahydroindolinone on further heating at 160 degrees C. The stereochemical outcome of the IMDAF cycloaddition has the side arm of the tethered vinyl group oriented exo with respect to the oxygen bridge. The resulting cycloadduct was used for the stereocontrolled installation of the remaining functionality present in the C-ring of the target molecules. Key features of the synthetic strategy include (1) a lithium hydroxide induced tandem hydrolysis/decarboxylation/elimination sequence to introduce the required pi-bond in the C-ring of (+/-)-lycoricidine, and (2) conversion of the initially formed Diels-Alder adduct into an aldehyde intermediate which then undergoes a stereospecific decarbonylation reaction mediated by Wilkinson's catalyst to set the trans-B-C ring junction of (+/-)-7-deoxypancratistatin.
Sigma-Aldrich
6-(5H)-Phenanthridinone, technical grade
Sigma-Aldrich
2-Iodobenzoic acid, 98%