Merck
  • In vitro scavenging activity for reactive oxygen species by N-substituted indole-2-carboxylic acid esters.
In vitro scavenging activity for reactive oxygen species by N-substituted indole-2-carboxylic acid esters.
Luminescence : the journal of biological and chemical luminescence (2007-05-02)
Irena Kruk, Hassan Y Aboul-Enein, Teresa Michalska, Krzysztof Lichszteld, Katarzyna Kubasik-Kladna, Sureyya Olgen
ABSTRACT
The hydroxyl radical (HO*)- and superoxide anion radical (O* (2))-scavenging activity, as well as the singlet oxygen ((1)O(2))-quenching property of N-substituted indole-2-carboxylic acid esters (INDs) were investigated by deoxyribose degradation assay, a chemiluminescence method and the electron spin resonance (ESR) spin-trapping technique. This novel group of compounds was developed as a search for cyclooxygenase-2 (COX-2)-selective enzyme inhibitors. The results obtained demonstrated that of the 16 compounds examined, five inhibited light emission from the superoxide anion radical (O* (2))-DMSO system by at least 60% at a concentration of 1 mmol/L, nine prevented the degradation of deoxyribose induced by the Fenton reaction system (range 3-78%) or scavenged hydroxyl radicals (HO*) directly (range 8-93%) and 14 showed the (1)O(2)-quenching effect (range 10-74%). These results indicate that majority of the indole esters tested possess the ability to scavenge O(-) (2) and HO radicals and to quench (1)O(2) directly, and consequently may be considered effective antioxidative agents.
Sigma-Aldrich
Indole-2-carboxylic acid, 98%