Merck
  • Preparation of optically active 2-aminobutanoic acid via optical resolution by replacing crystallization.
Preparation of optically active 2-aminobutanoic acid via optical resolution by replacing crystallization.
Bioscience, biotechnology, and biochemistry (2007-05-09)
Tatsuo Yajima, Yukiyo Aizawa, Mai Nishida, Yusuke Sakaguchi, Tadashi Shiraiwa
ABSTRACT
An attempt was made to use a simple procedure to obtain (R)- and (S)-2-aminobutanoic acids [(R)- and (S)-1] which are non-proteinogenic alpha-amino acids and are useful as chiral reagents in asymmetric syntheses. Compound (RS)-1 p-toluenesulfonate [(RS)-2], which is known to exist as a conglomerate, was optically resolved by replacing crystallization with (R)- and (S)-methionine p-toluenesulfonate [(R)- and (S)-3] as optically active co-solutes. When (S)-3 was employed as the co-solute, (R)-2 was preferentially crystallized from a supersaturated solution of (RS)-2 in 1-propanol, as was (S)-2 in the presence of (R)-3. (R)- and (S)-2 recrystallized from 1-propanol were treated with triethylamine in methanol to give (R)- and (S)-1 in optically pure forms.
Sigma-Aldrich
D-2-Aminobutyric acid, 98%
Sigma-Aldrich
DL-2-Aminobutyric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture