Merck
  • A novel compound, maltolyl p-coumarate, attenuates cognitive deficits and shows neuroprotective effects in vitro and in vivo dementia models.
A novel compound, maltolyl p-coumarate, attenuates cognitive deficits and shows neuroprotective effects in vitro and in vivo dementia models.
Journal of neuroscience research (2007-06-30)
Ki Young Shin, Geon Ho Lee, Cheol Hyoung Park, Hee Jin Kim, Soo-Hyun Park, Seonghan Kim, Hye-Sun Kim, Kwan-Sun Lee, Beom Young Won, Hyung Gun Lee, Jin-Ho Choi, Yoo-Hun Suh
ABSTRACT
To develop a novel and effective drug that could enhance cognitive function and neuroprotection, we newly synthesized maltolyl p-coumarate by the esterification of maltol and p-coumaric acid. In the present study, we investigated whether maltolyl p-coumarate could improve cognitive decline in scopolamine-injected rats and in amyloid beta peptide(1-42)-infused rats. Maltolyl p-coumarate was found to attenuate cognitive deficits in both rat models using passive avoidance test and to reduce apoptotic cell death observed in the hippocampus of the amyloid beta peptide(1-42)-infused rats. We also examined the neuroprotective effects of maltolyl p-coumarate in vitro using SH-SY5Y cells. Cells were pretreated with maltolyl p-coumarate, before exposed to amyloid beta peptide(1-42), glutamate or H2O2. We found that maltolyl p-coumarate significantly decreased apoptotic cell death and reduced reactive oxygen species, cytochrome c release, and caspase 3 activation. Taking these in vitro and in vivo results together, our study suggests that maltolyl p-coumarate is a potentially effective candidate against Alzheimer's disease that is characterized by wide spread neuronal death and progressive decline of cognitive function.
Sigma-Aldrich
Maltol, natural, FG
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG