Merck
  • FT-IR, FT-Raman spectra and ab initio HF, DFT vibrational analysis of p-chlorobenzoic acid.
FT-IR, FT-Raman spectra and ab initio HF, DFT vibrational analysis of p-chlorobenzoic acid.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-07-31)
N Sundaraganesan, B Anand, C Meganathan, B Dominic Joshua
ABSTRACT
The infrared, the Fourier transform infrared and Fourier transform Raman spectra of p-chlorobenzoic acid (p-CBA) has been recorded in the region 4000-600 cm(-1), 4000-400 cm(-1) and 4000-100 cm(-1), respectively. The optimized geometry, frequency and intensity of the vibrational bands of p-CBA were obtained by the ab initio HF and DFT (B3LYP) methods with complete relaxation in the potential energy surface using 6-311+G(d,p) basis set. The harmonic-vibrational frequencies were calculated and the scaled values have been compared with experimental FT-IR and FT-Raman spectra. The observed and the calculated frequencies are found to be in good agreement. The experimental spectra also coincide satisfactorily with those of theoretically constructed bar type spectrograms.
Sigma-Aldrich
4-Chlorobenzoic acid, 99%