Merck
  • Microbial metabolism of quinoline and related compounds. XI. Degradation of quinoline-4-carboxylic acid by Microbacterium sp. H2, Agrobacterium sp. 1B and Pimelobacter simplex 4B and 5B.
Microbial metabolism of quinoline and related compounds. XI. Degradation of quinoline-4-carboxylic acid by Microbacterium sp. H2, Agrobacterium sp. 1B and Pimelobacter simplex 4B and 5B.
Biological chemistry Hoppe-Seyler (1991-11-01)
M Schmidt, P Röger, F Lingens
ABSTRACT
From soil enrichment cultures four strains, using quinoline-4-carboxylic acid as sole source of energy and carbon, have been isolated. According to their physiological properties these bacteria have been identified as Microbacterium sp. designated H2, as Agrobacterium sp. designated 1b and Pimelobacter simplex designated 4B and 5B. Metabolites of the degradation pathway of quinoline-4-carboxylic acid have been isolated and identified. With Pimelobacter simplex 4B and 5B 2-oxo-1,2-dihydroquinoline-4-carboxylic acid and 8-hydroxycoumarin-4-carboxylic acid were isolated. The Agrobacterium strain accumulated 2-oxo-1,2-dihydroquinoline-4-carboxylic acid and 2-oxo-1,2,3,4-tetrahydroquinoline-4-carboxylic acid in the media during growth; with Microbacterium sp. H2 we only found 8-hydroxycoumarin-4-carboxylic acid. With mutants of Microbacterium sp. H2 which were induced with N-methyl-N'-nitro-N-nitrosoguanidine we found 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 8-hydroxy-coumarin-4-carboxylic acid and 2,3-dihydroxyphenyl-succinic acid.
Sigma-Aldrich
4-Quinolinecarboxylic acid, 97%