Merck
  • Simultaneous analysis of lysine, Nepsilon-carboxymethyllysine and lysinoalanine from proteins.
Simultaneous analysis of lysine, Nepsilon-carboxymethyllysine and lysinoalanine from proteins.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2007-11-13)
Lourdes Bosch, Maria Luz Sanz, Antonia Montilla, Amparo Alegría, Rosaura Farré, María Dolores del Castillo
ABSTRACT
Protein quality was assayed by simultaneous measurement of lysine (Lys), carboxymethyllysine (CML) and lysinoalanine (LAL). GC-FID analysis of N-tert-butyl dimethylsilyl (tBDMSi) derivatives of these amino acids was undertaken. tBDMSi derivates were separated on a CP-SIL 5CB commercially fused silica capillary column (25 m x 0.25 mm i.d., 0.25 microm film thickness) employing a thermal gradient programmed from 200 to 300 degrees C. The identity of tBDMSi derivatives of Lys, CML and LAL was established by GC-MS while FID detection was employed for quantification. Analytical parameters such as linearity (lysine 350-4200 microM, LAL 3-81 microM, CML 16-172 microM), precision (1-13% variation coefficients), accuracy (85-108% average recovery) and limits of detection (lysine 0.4 mg/100 g protein, LAL 5.0 mg/100 g protein, CML 3.4 mg/100 g protein) and quantification (lysine 1.4 mg/100g protein, LAL 15.2 mg/100 g protein, CML 11.2 mg/100 g protein) were determined for validation of the analytical approach. Model systems and real foods have been studied. Kinetic of CML formation from different food proteins (BSA, soy protein, casein and gluten) was performed employing model systems. Carboxymethylation rate depended on the source of protein. Maillard reaction progressed to advanced stages damaging the protein quality of stored infant foods, soy drinks, boiled eggs and dry powdered crepes. CML values ranged from 62 to 440 mg/100 g protein were measured. LAL was also formed during boiling eggs (21-68 mg/100g protein) indicating additional damage by crosslinking reaction. In agreement, lysine content was affected by both food processing and storage.
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%