Merck
  • 1H NMR investigations of inclusion complexes between beta-cyclodextrin and 1-hexadecanol.
1H NMR investigations of inclusion complexes between beta-cyclodextrin and 1-hexadecanol.
Current drug discovery technologies (2007-11-30)
Huiming Jiang, Shufen Zhang, Qinglin Guan, Chang Chen, Fan Gao, Yang Zhang
ABSTRACT
The inclusion complex between beta-CD and 1-hexadecanol is synthesized and identified via (1)H NMR spectrum. The possible conformation of the inclusion complex is figured out. Via MM2 calculations, the possibility of complexation is verified.
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cetyl alcohol, analytical standard