Merck
  • New radiopaque acrylic bone cement. II. Acrylic bone cement with bromine-containing monomer.
New radiopaque acrylic bone cement. II. Acrylic bone cement with bromine-containing monomer.
Journal of materials science. Materials in medicine (2008-01-17)
M C Rusu, I C Ichim, M Popa, M Rusu
ABSTRACT
Bromine-containing methacrylate, 2-(2-bromopropionyloxy) ethyl methacrylate (BPEM), had been used in the formulation of acrylic radiopaque cements. The effect of this monomer incorporated into the liquid phase of acrylic bone cement, on the curing parameters, thermal properties, water absorption, density, compression tests and radiopacity was studied. A decrease of maximum temperature and an increase of the setting time were observed with the addition of the bromine-containing monomer in the radiolucent cement composition. Adding BPEM in radiolucent acrylic bone cements composition results in the decrease of glass transition temperature and increase of its thermal stability. Acrylic bone cements modified with bromine-containing comonomer are characterized by polymerization shrinkage lower than the radiolucent cement. Addition of bromine-containing comonomer in radiolucent acrylic bone cement composition determines the increase of compressive strength. Acrylic bone cements modified with bromine-containing comonomer proved to be radiopaque.
Sigma-Aldrich
Ethyl methacrylate, contains 15-20 ppm monomethyl ether hydroquinone as inhibitor, 99%
Pricing and availability is not currently available.