Merck
  • A sensitive rutin assay using a simple probe manganese sulfate based on its novel resonance light scattering decrease phenomenon.
A sensitive rutin assay using a simple probe manganese sulfate based on its novel resonance light scattering decrease phenomenon.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-02-20)
Zhanguang Chen, Li Zhu, Junhui Chen, Jinbin Liu, Yali Han
ABSTRACT
A novel resonance light scattering (RLS) decrease method was developed to determine rutin with a simple probe manganese sulfate. At pH 7.5, the strong RLS intensity of manganese sulfate was remarkably decreased by the addition of rutin with the maximum peak located at 389.0 nm. Under the optimum conditions, a good linear relationship between the changes of RLS intensities of manganese sulfate with and without rutin and the concentrations of rutin was obtained over the range of 0.49-24.4 microg ml(-1) and a low detection limit (3sigma) 0.42 microg ml(-1) was achieved in the mean time. Based on this approach, a novel method for quantitative analysis of rutin is proposed in this paper. The method proposed was also applied successfully to the determination of rutin in commercial pharmaceutical preparations of compound rutin tablets and human urine samples. The assay is sensitive, rapid, inexpensive, practical and relatively free from interference generated by coexisting substances.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%