Merck
Microbial metabolism of 1-aminoanthracene by Beauveria bassiana.
Bioorganic & medicinal chemistry (2008-04-02)
Jixun Zhan, A A Leslie Gunatilaka
ABSTRACT
The carcinogen and mutagen, 1-aminoanthracene, was efficiently metabolized by the fungal strain Beauveria bassiana ATCC 7159 to yield three new metabolites identified as 1-acetamido-5-[(4'-O-methyl-beta-d-glucopyranosyl)oxy]anthracene, 1-acetamido-8-[(4'-O-methyl-beta-d-glucopyranosyl)oxy]anthraquinone, and 1-acetamido-6-[(4'-O-methyl-beta-d-glucopyranosyl)oxy]anthraquinone, together with 1-acetamidoanthracene and 1-acetamidoanthraquinone. Formation of these metabolites suggests that the metabolic pathways of 1-aminoanthracene in B. bassiana ATCC 7159 involve acetylation, oxidation, hydroxylation, and O-methylglucosylation.
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%