Merck
  • Evaluation of the users value of salts against apple scab and powdery mildew for the integrated fruit production.
Evaluation of the users value of salts against apple scab and powdery mildew for the integrated fruit production.
Communications in agricultural and applied biological sciences (2008-04-10)
P Creemers, S Van Laer, A Van Mechelen, B Vorstermans, K Hauke
ABSTRACT
As new fungicides are mainly unisite action fungicides, the problem of fungicide resistance development is becoming more important every year. Combining chemical fungicides, which is the best anti-resistance strategy, is not always possible or recommended in the case when the number of available chemical fungicides are limited or a reduction in fungicide use is asked for. Therefore the use of salts as an anti-resistance strategy was looked upon. The salts evaluated were K(HCO3), KH2PO3, KHPO4 and K2SiO3. When using these salts as an anti-resistance strategy the efficacy obtained when spraying the compounds alone was often to low to be used in rotation with chemical fungicides. Only with K(HCO3)2 a good efficacy can be observed in some years. The variation in efficacy with K(HCO3)2 observed is higher for powdery mildew. Chitosan was also included in the trials against powdery mildew, however chitosan had no effect on the infestation.
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules