Merck
Selector for structural isomers of neutral molecules.
Physical review letters (2008-06-04)
Frank Filsinger, Undine Erlekam, Gert von Helden, Jochen Küpper, Gerard Meijer
ABSTRACT
We have selected and spatially separated the two conformers of 3-aminophenol (C(6)H(7)NO) present in a molecular beam. Analogous to the separation of ions based on their mass-to-charge ratios in a quadrupole mass filter, the neutral conformers are separated based on their different mass-to-dipole-moment ratios in an ac electric quadrupole selector. For a given ac frequency, the individual conformers experience different focusing forces, resulting in different transmissions through the selector. These experiments demonstrate that conformer-selected samples of large molecules can be prepared, offering new possibilities for the study of gas-phase biomolecules.
Sigma-Aldrich
3-Aminophenol, 98%
Supelco
3-Aminophenol, PESTANAL®, analytical standard