Merck
[Brain targeting of gastrodin nasal in situ gel].
Sichuan da xue xue bao. Yi xue ban = Journal of Sichuan University. Medical science edition (2008-06-26)
Zheng Cai, Shi-Xiang Hou, Zhao-Xiang Yang, Xiang-Rong Song, Qiu-Hong Chen, Yuan-Bo Li, Bin-Bin Zhao
ABSTRACT
To investigate the biodistribution of gastrodin ion-activated nasal in situ gel in rat blood and brain tissues and to evaluate its brain targeting. Intravenous administration of gastrodin solution or intranasal administration of gastrodin nasal in situ gel were given to 32 rats, respectively. The concentrations of gastrodin in the plasma and gastrodigenin in the brain tissues of the rats were determined by HPLC. The intranasal administration of the in situ gel of gastrodin produced more significant brain targeting effect than the intravencus administration of gstrodin solution (P < 0.01). The area under curve (AUC) of cerebrum, cerebellum and olfactory bulb increased by 1.16, 0.77 and 3.34 times, with 2.66. 2.18 and 5.34 brain targeting indexes (BTI), respectively. The mean residence time (MRT) increased by nearly four-folds. Gastrodin nasal in situ gel can improve the brain targeting of gastrodin and slow its release.
Sigma-Aldrich
4-Hydroxybenzyl alcohol, 99%
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG