Merck
  • Ozonolysis of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal: kinetics and mechanisms.
Ozonolysis of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal: kinetics and mechanisms.
The journal of physical chemistry. A (2008-07-01)
Tadeu Leonardo, Edilson Clemente da Silva, Graciela Arbilla
ABSTRACT
A combined density functional theory and transition-state theory study of the mechanisms and reaction coefficients of gas-phase ozonolysis of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal is presented. The geometries, energies, and harmonic vibrational frequencies of each stationary point were determined by B3LYP/6-31(d,p), MPW1K/cc-pVDZ, and BH&HLYP/cc-pVDZ methods. According to the calculations, the ozone 6-methyl-5-hepten-2-one reaction is faster than the ozone 6-hydroxy-4-methyl-4-hexenal reaction, but both are slower than the ozone geraniol-trans reaction. By using the BH&HLYP/cc-pVDZ data, a global rate coefficient of 5.9 x 10(-16) cm(3) molecule(-1) s(-1) was calculated, corresponding to the sum of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal reactions with the ozone. These results are in good agreement with the experimental studies.
Sigma-Aldrich
6-Methyl-5-hepten-2-one, 99%
Sigma-Aldrich
6-Methyl-5-hepten-2-one, ≥98%, FCC, FG
Sigma-Aldrich
6-Methyl-5-hepten-2-one, natural, ≥98%, FCC
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®