Merck
  • Catalytic ozonation of p-chlorobenzoic acid by activated carbon and nickel supported activated carbon prepared from petroleum coke.
Catalytic ozonation of p-chlorobenzoic acid by activated carbon and nickel supported activated carbon prepared from petroleum coke.
Journal of hazardous materials (2008-08-01)
Xukai Li, Qiuyun Zhang, Lili Tang, Ping Lu, Fengqiang Sun, Laisheng Li
ABSTRACT
The aim of this research was to investigate catalytic activity of petroleum coke, activated carbon (AC) prepared from this material, Ni supported catalyst on activated carbon (Ni/AC) in the ozonation of aqueous phase p-chlorobenzoic acid (p-CBA). Activated carbon and Ni/AC catalyst were characterized by XRD and SEM. The presence of petroleum coke did not improve the degradation of p-CBA compared to ozonation alone, but it was advantageous for p-CBA mineralization (total organic carbon, TOC, reduction), indicating the generation of highly oxidant species (*OH) in the medium. The presence of either activated carbon or Ni/AC considerably improves TOC removal during p-CBA ozonation. Ni/AC catalyst shows the better catalytic activity and stability based on five repeated tests during p-CBA ozonation. During the ozonation (50 mg/h ozone flow rate) of a 10 mg/L p-CBA (pH 4.31), it can be more mineralized in the presence of Ni/AC catalyst (5.0 g/L), TOC removal rate is over 60% in 60 min, 43% using activated carbon as catalyst, only 30% with ozonation alone.
Sigma-Aldrich
4-Chlorobenzoic acid, 99%