Merck
Kinetic investigation of erucamide synthesis using fatty acid and urea.
Journal of oleo science (2008-08-08)
Neeraj Praphulla Awasthi, Santosh K Upadhayay, R P Singh
ABSTRACT
Fatty acid amides like erucamide are mainly used for lubrication and as slip agent to decrease friction in polymer and plastic industry. Erucamide is normally synthesized by ammonolysis of triglycerides or fatty acids at 200 degrees C and at high pressure (345-690 kPa.). However using urea in place of ammonia the economic synthesis of erucamide is possible at atmospheric pressure at approx 190 degrees C. In present investigation, the kinetics of synthesis of erucamide by ammonolysis of erucic acid has been investigated. The optimum conditions for the synthesis of erucamide have also been determined. 1:4 molar ratio of erucic acid to urea, 190 degrees C temperature and catalyst [P2O5 with (NH4)2H PO4, {(1:1) w/w }] concentration 3% (by wt. of erucic acid) were the optimum condition for synthesis of erucamide from erucic acid and can obtain a maximum yield of 92% of pure erucamide. Some other catalysts as titanium-iso -propoxide, phosphorus pent oxide were also tried but these catalysts were not economical.
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
cis-13-Docosenoamide