Merck
  • Dipole effects on cation-pi interactions: absolute bond dissociation energies of complexes of alkali metal cations to N-methylaniline and N,N-dimethylaniline.
Dipole effects on cation-pi interactions: absolute bond dissociation energies of complexes of alkali metal cations to N-methylaniline and N,N-dimethylaniline.
The journal of physical chemistry. A (2008-08-14)
Nuwan Hallowita, Damon R Carl, P B Armentrout, M T Rodgers
ABSTRACT
Threshold collision-induced dissociation of M (+)( nMA) x with Xe is studied using guided ion beam mass spectrometry, where nMA = N-methylaniline and N, N-dimethylaniline and x = 1 and 2. M (+) includes the following alkali metal cations: Li (+), Na (+), K (+), Rb (+), and Cs (+). In all cases, the primary dissociation pathway corresponds to the endothermic loss of an intact nMA ligand. The primary cross section thresholds are interpreted to yield 0 and 298 K bond dissociation energies (BDEs) for ( nMA) x-1 M (+)-( nMA) after accounting for the effects of multiple ion-neutral collisions, the internal and kinetic energy distributions of the reactants, and the dissociation lifetimes. Density functional theory calculations at the B3LYP/6-31G* level of theory are used to determine the structures of these complexes, which are also used in single-point calculations at the MP2(full)/6-311+G(2d,2p) level to determine theoretical BDEs. The results of these studies are compared to previous studies of the analogous M (+)(aniline) x complexes to examine the effects of methylation of the amino group on the binding interactions. Comparisons are also made to a wide variety of cation-pi complexes previously studied to elucidate the contributions that ion-dipole, ion-induced-dipole, and ion-quadrupole interactions make to the overall binding.
Sigma-Aldrich
N,N-Dimethylaniline, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Sigma-Aldrich
N-Methylaniline, 98%
Sigma-Aldrich
N-Methylaniline, ≥99%
Supelco
N-Methylaniline, analytical standard