- Salicylanilides: selective inhibitors of interleukin-12p40 production.
Salicylanilides: selective inhibitors of interleukin-12p40 production.
Bioorganic & medicinal chemistry (2008-08-22)
Michael E Brown, Jeffrey N Fitzner, Tracey Stevens, Wilson Chin, Clifford D Wright, Jim P Boyce
PMID18715785
ABSTRACT
Interleukin (IL)-12p40, a subunit component of both IL-12 and IL-23, is being widely studied for its role in inflammatory disease. As part of an effort to profile cellular signaling pathways across different cell types, we report salicylanilide inhibitors of IL-12p40 production in stimulated dendritic cells. Based on a hypothesis that a desirable therapeutic profile is one that could block IL-12p40 but not IL-6 production, we engaged in directed analoging. This resulted in salicylanilides with similar IL-12p40 related potency but enhanced selectivity relative to IL-6 production.