Merck
  • Chloroalkylsulfonate ionic liquids by ring opening of sultones with organic chloride salts.
Chloroalkylsulfonate ionic liquids by ring opening of sultones with organic chloride salts.
Chemical communications (Cambridge, England) (2008-08-30)
Natalia Paape, Wei Wei, Andreas Bösmann, Claudia Kolbeck, Florian Maier, Hans-Peter Steinrück, Peter Wasserscheid, Peter Steffen Schulz
ABSTRACT
An efficient route to prepare ionic liquids with chloroalkylsulfonate anions is presented; the synthesis proceeds in a one-step ring-opening reaction of sultones with an organic chloride salt and provides a very attractive access to new anion functionalised ionic liquids.