Merck
  • Solubilisation of griseofulvin in aqueous micellar solutions of diblock copolymers of ethylene oxide and 1,2-butylene oxide with lengthy B-blocks.
Solubilisation of griseofulvin in aqueous micellar solutions of diblock copolymers of ethylene oxide and 1,2-butylene oxide with lengthy B-blocks.
International journal of pharmaceutics (2008-12-02)
Maria Elenir N P Ribeiro, Igor Marques Cavalcante, Nágila M P S Ricardo, Shao-Min Mai, David Attwood, Stephen G Yeates, Colin Booth
ABSTRACT
The influence of hydrophobic-block length on solubilisation capacity was examined for micelles of E(m)B(n) copolymers (E=oxyethylene, B=oxybutylene, subscripts denote number-average block lengths in repeat units) with B-block lengths in the range of 30-76 and with E-blocks of sufficient length to ensure the formation of spherical micelles. Griseofulvin was used as a model poorly-water-soluble drug known to be almost exclusively solubilised in the micellar core. Combination of solubilisation data with those of a previous study has shown that the amount of drug solubilised per gram of hydrophobe is essentially independent of B-block length when this exceeds about 15 B units, suggesting that core size is not a major influence on solubilisation.
Sigma-Aldrich
(R)-(+)-1,2-Epoxybutane, 98%
Sigma-Aldrich
(S)-(−)-1,2-Epoxybutane, 98%
Sigma-Aldrich
1,2-Epoxybutane, 99%