Merck
  • Vanadium-induced nucleophilic IPSO substitutions in a coordinated tetrachlorosemiquinone ring: formation of the chloranilate anion as a bridging ligand.
Vanadium-induced nucleophilic IPSO substitutions in a coordinated tetrachlorosemiquinone ring: formation of the chloranilate anion as a bridging ligand.
Inorganic chemistry (2009-01-09)
Pabitra Baran Chatterjee, Kisholoy Bhattacharya, Nabanita Kundu, Ki-Young Choi, Rodolphe Clérac, Muktimoy Chaudhury
ABSTRACT
In basic media, the coordinated semiquinone radical in the spin-coupled [(bipy)ClV(IV)O(TCSQ)] 1 (HTCSQ = tetrachlorosemiquinone) undergoes nucleophilic ipso substitution (OH- for Cl-) to generate the chloranilate anion (CA(2-)) that bridges the vanadium(IV) centers, forming a binuclear compound [(bipy)ClV(IV)O(CA)OV(IV)Cl(bipy)] 2.
Sigma-Aldrich
Chloranilic acid, ≥98%