Merck
  • Novel norhumulene and xeniaphyllane-derived terpenoids from a formosan soft coral Sinularia gibberosa.
Novel norhumulene and xeniaphyllane-derived terpenoids from a formosan soft coral Sinularia gibberosa.
Chemical & pharmaceutical bulletin (2009-02-03)
Shin-Pin Chen, Jui-Hsin Su, Hsiao-Chien Yeh, Atallah Fouad Ahmed, Chang-Feng Dai, Yang-Chang Wu, Jyh-Horng Sheu
ABSTRACT
A novel skeletal norhumulene (1) and six xeniaphyllane-derived compounds, including norditerpenoids (2, 3) and diterpenoids (4-7), were isolated from the EtOAc extract of the Formosan soft coral Sinularia gibberosa. Their structures were elucidated by spectroscopic analysis. In vitro cytotoxic evaluation of the above metabolites revealed that 2 and 4 showed weak cytotoxicity toward a few cancer cell lines.