Merck
Novel bUT-B2 urea transporter isoform is constitutively activated.
American journal of physiology. Regulatory, integrative and comparative physiology (2009-05-29)
P Tickle, A Thistlethwaite, C P Smith, G S Stewart
ABSTRACT
Our previous studies have detailed a novel facilitative UT-B urea transporter isoform, bUT-B2. Despite the existence of mouse and human orthologs, the functional characteristics of UT-B2 remain undefined. In this report, we produced a stable MDCK cell line that expressed bUT-B2 protein and investigated the transepithelial urea flux across cultured cell monolayers. We observed a large basal urea flux that was significantly reduced by known inhibitors of facilitative urea transporters; 1,3 dimethylurea (P < 0.001, n = 17), thionicotinamide (P < 0.05, n = 11), and phloretin (P < 0.05, n = 9). Pre-exposure for 1 h to the antidiuretic hormone vasopressin had no effect on bUT-B2-mediated urea transport (NS, n = 3). Acute vasopressin exposure for up to 30 min also failed to elicit any transient response (NS, n = 9). Further investigation confirmed that bUT-B2 function was not affected by alteration of intracellular cAMP (NS, n = 4), intracellular calcium (NS, n = 3), or protein kinase activity (NS, n = 4). Finally, immunoblot data suggested a possible role for glycosylation in regulating bUT-B2 function. In conclusion, this study showed that bUT-B2-mediated transepithelial urea transport was constitutively activated and unaffected by known regulators of renal UT-A urea transporters.
Sigma-Aldrich
N,N′-Dimethylurea, (sym.), ≥95.0% (HPLC), technical
Supelco
N,N′-Dimethylurea, PESTANAL®, analytical standard
Sigma-Aldrich
N,N′-Dimethylurea, (sym.), ≥99% (from N)