Merck
  • Effect of low and zero magnetic field on the hyperpolarization lifetime in parahydrogenated perdeuterated molecules.
Effect of low and zero magnetic field on the hyperpolarization lifetime in parahydrogenated perdeuterated molecules.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2009-06-19)
Francesca Reineri, Daniela Santelia, Roberto Gobetto, Silvio Aime
ABSTRACT
Hyperpolarization can be kept for times longer than T(1) if it is maintained in a singlet state, from which transitions are not allowed. Another, more direct, way to slower the relaxation process consists in the use of perdeuterated molecules. Here both methods have been applied and the hyperpolarization (induced by para-H(2)) decay rate has been measured at two different magnetic fields: earth field and zero field. While relaxation is very slow at earth field, it becomes faster at zero field: this rather unexpected finding has been explained on the basis of isotropic mixing between (1)H and (2)H.
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG