Merck
  • Synthesis and characterization of toluene-3,4-dithiolatoantimony(III) derivatives with some oxygen and/or sulphur donor ligands.
Synthesis and characterization of toluene-3,4-dithiolatoantimony(III) derivatives with some oxygen and/or sulphur donor ligands.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-06-30)
H P S Chauhan, Sumit Bhatiya, Abhilasha Bakshi
ABSTRACT
Replacement reactions of toluene-3,4-dithiolatoantimony(III) chloride with oxygen and/or sulphur donor ligands like benzoic acid, thiobenzoic acid, thioacetic acid, phenol, thiophenol, sodium salicylate and thio glycolic acid in 1:1 molar ratio as well as disodium oxalate in 2:1 molar ratio in refluxing anhydrous benzene yielded toluene-3,4-dithiolatoantimony(III) mono oxo and/or thio carboxylic or phenolic derivatives of the general formula {R=OOCC(6)H(5), SOCC(6)H(5), SOCCH(3), OC(6)H(5), SC(6)H(5), OOCC(6)H(4)(OH) and SCH(2)COOH} and These newly synthesized derivatives are yellow and brown solids/liquids and are soluble in common organic solvents like benzene, chloroform, dichloromethane, etc. These derivatives have been characterized by melting point determination, molecular weight determination, elemental analysis (C, H, S and Sb), spectral {UV, IR and NMR ((1)H and (13)C)} and thermal (TGA, DTA and DSC) studies.
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Potassium thioacetate, 98%
Sigma-Aldrich
Thioacetic acid, 96%