Merck
  • Solid-phase synthesis and DNA binding studies of dichloroplatinum(ii) conjugates of dicarba analogues of octreotide as new anticancer drugs.
Solid-phase synthesis and DNA binding studies of dichloroplatinum(ii) conjugates of dicarba analogues of octreotide as new anticancer drugs.
Chemical communications (Cambridge, England) (2009-07-31)
Flavia Barragán, Virtudes Moreno, Vicente Marchán
ABSTRACT
The first dichloroplatinum(ii) conjugates of dicarba analogues of octreotide, which is expected to act as a 'tumour-targeting device', have been efficiently synthesized following a stepwise solid-phase approach; these compounds emulate the mechanism of cisplatin since they form a 1,2-intrastrand cross-link with two consecutive guanines of an oligonucleotide.
Product Description
Supelco
Dicamba, PESTANAL®, analytical standard
Sigma-Aldrich
Dicamba, suitable for plant cell culture, BioReagent