Merck
  • Asymmetric histidine-catalyzed cross-aldol reactions of enolizable aldehydes: access to defined configured quaternary stereogenic centers.
Asymmetric histidine-catalyzed cross-aldol reactions of enolizable aldehydes: access to defined configured quaternary stereogenic centers.
Journal of the American Chemical Society (2009-11-03)
Morris Markert, Ulf Scheffler, Rainer Mahrwald
ABSTRACT
A histidine-catalyzed asymmetric direct cross-aldol reaction of enolizable aldehydes is described. In contrast to proline, histidine is able to clearly differentiate the reactivity of various aldehydes. In addition, this approach provides access to syn-configured beta-hydroxyaldehydes. Thus, by application of this new methodology, defined-configuration quaternary stereocenters can be constructed with ease. The utility of this method is demonstrated in several total syntheses of branched-chain carbohydrates.
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
D-(−)-Lyxose, 99%