Merck
  • Kinetic characterization and comparison of various protein crosslinking reagents for matrix modification.
Kinetic characterization and comparison of various protein crosslinking reagents for matrix modification.
Journal of materials science. Materials in medicine (2010-01-14)
Paul Slusarewicz, Keng Zhu, Tom Hedman
ABSTRACT
We have characterized the relative efficacies of a number of protein crosslinking agents that have the potential for use in the crosslinking of proteinaceous matrices both in vitro and in vivo. The crosslinkers tested were; L: -threose (LT), Genipin (GP), Methylglyoxal (MG), 1-ethyl-3-(3-dimethylaminopropyl) carbodiimide hydrochloride (EDC), proanthrocyanidin (PA) and glutaraldehyde (GA). The relative effectiveness of the crosslinkers with regard to their saturating concentrations was: GA > PA > EDC > MG = GP > LT. Most of the crosslinkers displayed a pH dependence and were more effective at more alkaline pH. At optimal pH and saturating conditions, the relative reaction rates of the crosslinkers were: PA = GA > EDC > GP > MG > LT.