Merck
  • A simple and rapid technique for recovery of 99mTc from low specific activity (n,gamma)99Mo based on solid-liquid extraction and column chromatography methodologies.
A simple and rapid technique for recovery of 99mTc from low specific activity (n,gamma)99Mo based on solid-liquid extraction and column chromatography methodologies.
Nuclear medicine and biology (2010-02-04)
Sankha Chattopadhyay, Sujata Saha Das, Luna Barua
ABSTRACT
A simple and inexpensive method has been developed for the separation of (99m)Tc from (99)Mo produced from the neutron activation of (98)Mo by (98)Mo(n,gamma)(99)Mo nuclear reaction. The recovery of (99m)Tc was performed by solid-liquid extraction based on alumina column chromatography. The overall radiochemical yield for the complete separation of (99m)Tc was 85-97% (n=5). The separated Na[(99m)Tc]TcO(4) was of high radionuclidic, radiochemical and chemical purities. The method can be adopted for routine processing and use of (99m)Tc in radiopharmacy operations.
Sigma-Aldrich
Sodium molybdate, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate, ≥98%