Merck
  • Allergic contact dermatitis from ethylhexyl salicylate and other salicylates.
Allergic contact dermatitis from ethylhexyl salicylate and other salicylates.
Dermatitis : contact, atopic, occupational, drug (2010-03-18)
Charlotte Gotthard Mortz, Henrik Thormann, An Goossens, Klaus Ejner Andersen
ABSTRACT
Allergic contact dermatitis (ACD) from salicylates present in topical products is uncommon. Most publications about ACD from salicylates are case reports describing only a few patients. Cross-reactivity between salicylates is not commonly reported. This article describes allergic contact dermatitis from ethylhexyl salicylate used as an ultraviolet filter and fragrance compound and reviews the published literature on contact allergy to salicylates.
Product Description
Supelco
2-Ethylhexyl salicylate, analytical standard
Sigma-Aldrich
2-Ethylhexyl salicylate, ≥99%
Sigma-Aldrich
2-Ethylhexyl salicylate, 99%
Supelco
Octisalate, Pharmaceutical Secondary Standard; Certified Reference Material