Merck
  • The effectiveness of benzydamine hydrochloride spraying on the endotracheal tube cuff or oral mucosa for postoperative sore throat.
The effectiveness of benzydamine hydrochloride spraying on the endotracheal tube cuff or oral mucosa for postoperative sore throat.
Anesthesia and analgesia (2010-06-29)
Yuan-Shiou Huang, Nan-Kai Hung, Meei-Shyuan Lee, Chang-Po Kuo, Jyh-Cherng Yu, Go-Shine Huang, Chen-Hwan Cherng, Chih-Shung Wong, Chi-Hong Chu, Ching-Tang Wu
ABSTRACT
The etiology of postoperative sore throat (POST) is considered to be the result of laryngoscopy, intubation damage, or inflated cuff compression of the tracheal mucosa. In this study, we compared the effectiveness in alleviating POST using different approaches to benzydamine hydrochloride (BH) administration by spraying the endotracheal tube (ET) cuff or the oropharyngeal cavity, or both. Three hundred eighty patients were included in this prospective and double-blind study, which was randomized into 4 groups: group A, oropharyngeal cavity spray of BH, and distilled water on the ET cuff; group B, both the oropharyngeal cavity and the ET cuff received BH spray; group C, the ET cuff received BH spray, and the oropharyngeal cavity received distilled water; and group D, distilled water sprayed on both the ET tube and into the oropharyngeal cavity. The patients were examined for sore throat (none, mild, moderate, severe) at 0, 2, 4, and 24 hours postextubation. The incidence of POST was 23.2%, 13.8%, 14.7%, and 40.4% in groups A, B, C, and D, respectively. POST occurred significantly less frequently in groups B and C compared with group D (odds ratio: 0.36; 95% confidence interval: 0.21-0.60; P < 0.05). However, there was no significant difference between groups A and D (odds ratio: 0.62; 95% confidence interval: 0.38-1.01). Moreover, there was no significant interaction between spraying BH over the oropharyngeal cavity and the ET cuff on the incidence of POST (P = 0.088). The severity of POST was significantly more intense in group D compared with groups B and C (P < 0.001). Group B had a significantly higher incidence of local numbness, burning, and/or stinging sensation compared with patients in group D (P < 0.05). This study indicates that spraying BH on the ET cuff decreases the incidence and severity of POST without increased BH-related adverse effects.
Product Description
Supelco
Benzydamine hydrochloride, analytical standard