Merck
  • Gamma- and delta-tocotrienols inhibit skin melanin synthesis by suppressing constitutive and UV-induced tyrosinase activation.
Gamma- and delta-tocotrienols inhibit skin melanin synthesis by suppressing constitutive and UV-induced tyrosinase activation.
Pigment cell & melanoma research (2010-07-09)
W N Yap, N Zaiden, C H Xu, A Chen, S Ong, V Teo, Y L Yap
Product Description
Supelco
δ-Tocotrienol, analytical standard
Sigma-Aldrich
D-δ-Tocotrienol, from rice, ≥65% (GC)