Merck
  • Superior discriminating value of ACTH-stimulated serum 21-deoxycortisol in identifying heterozygote carriers for 21-hydroxylase deficiency.
Superior discriminating value of ACTH-stimulated serum 21-deoxycortisol in identifying heterozygote carriers for 21-hydroxylase deficiency.
Clinical endocrinology (2010-09-18)
Flávia A Costa-Barbosa, Vânia F Tonetto-Fernandes, Valdemir M Carvalho, Odete H Nakamura, Vivian Moura, Tânia A S S Bachega, José G H Vieira, Claudio E Kater
ABSTRACT
Congenital adrenal hyperplasia caused by classic 21-hydroxylase deficiency (21OHD) is an autosomal recessive disorder with a high prevalence of asymptomatic heterozygote carriers (HTZ) in the general population, making case detection desirable by routine methodology. HTZ for classic and nonclassic (NC) forms have basal and ACTH-stimulated values of 17-hydroxyprogesterone (17OHP) that fail to discriminate them from the general population. 21-Deoxycortisol (21DF), an 11-hydroxylated derivative of 17OHP, is an alternative approach to identify 21OHD HTZ. To determine the discriminating value of basal and ACTH-stimulated serum levels of 21DF in comparison with 17OHP in a population of HTZ for 21OHD (n = 60), as well as in NC patients (n = 16) and in genotypically normal control subjects (CS, n = 30), using fourth generation tandem mass spectrometry after HPLC separation (LC-MS/MS). Basal 21DF levels were not different between HTZ and CS, but stimulated values were increased in the former and virtually nonresponsive in CS. Only 17·7% of the ACTH-stimulated 21DF levels overlapped with CS, when compared to 46·8% for 17OHP. For 100% specificity, the sensitivities achieved for ACTH-stimulated 21DF, 17OHP and the quotient [(21DF + 17OHP)/F] were 82·3%, 53·2% and 87%, using cut-offs of 40, 300 ng/dl and 46 (unitless), respectively. Similar to 17OHP, ACTH-stimulated 21DF levels did not overlap between HTZ and NC patients. A positive and highly significant correlation (r = 0·846; P < 0·001) was observed between 21DF and 17OHP pairs of values from NC and HTZ. This study confirms the superiority of ACTH-stimulated 21DF, when compared to 17OHP, both measured by LC-MS/MS, in identifying carriers for 21OHD. Serum 21DF is a useful tool in genetic counselling to screen carriers among relatives in families with affected subjects, giving support to molecular results.
Sigma-Aldrich
21-Deoxycortisol
Supelco
11-Deoxycortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
21-Deoxycortisol solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reichstein′s substance S, ≥98%