Merck
  • Determination of carbamazepine and its impurities iminostilbene and iminodibenzyl in solid dosage form by column high-performance liquid chromatography.
Determination of carbamazepine and its impurities iminostilbene and iminodibenzyl in solid dosage form by column high-performance liquid chromatography.
Journal of AOAC International (2010-10-07)
Predrag Lj Dzodić, Ljiljana J Zivanović, Ana D Protić, Mira L Zećević, Biljana M Jocić
ABSTRACT
An accurate and precise RP-HPLC method was developed and validated for the determination of carbamazepine and its impurities iminostilbene and iminodibenzyl in a tablet formulation with fluphenazine as an internal standard. Buffer-methanol (50 + 50, v/v) was used as the mobile phase. During validation, specificity, linearity, precision, accuracy, LOD, LOQ, and robustness of the method were tested. The method was proven to be specific against placebo interference. Linearity was evaluated over the concentration range of 100-500, 0.05-0.25, and 0.1-0.5 microg/mL, and the r values were 0.9994, 0.9997, and 0.9979 for carbamazepine, iminostilbene, and iminodibenzyl, respectively. Intraday precision of the method was good, and RSD was below 2% for all analytes. The accuracy of the method ranged from 100.69 to 102.10, 99.76 to 102.66, and 99.26 to 100.08% for carbamazepine, iminostilbene, and iminodibenzyl, respectively. LOD was 0.0125, 0.025, and 0.05 microg/mL and LOQ was 0.05, 0.05, and 0.1 microg/mL for carbamazepine, iminostilbene, and iminodibenzyl, respectiviely. Robustness of the method was proven by using a chemometric approach. The method was successfully applied to the analysis of commercially available carbamazepine tablets and showed good repeatability, with RSD below 2%.
Product Description
Supelco
Iminostilbene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dibenzylamine, 97%
Sigma-Aldrich
5H-Dibenz[b,f]azepine, 97%