Merck
  • Chemical identity of a rotting animal-like odor emitted from the inflorescence of the titan arum (Amorphophallus titanum).
Chemical identity of a rotting animal-like odor emitted from the inflorescence of the titan arum (Amorphophallus titanum).
Bioscience, biotechnology, and biochemistry (2010-12-15)
Mika Shirasu, Kouki Fujioka, Satoshi Kakishima, Shunji Nagai, Yasuko Tomizawa, Hirokazu Tsukaya, Jin Murata, Yoshinobu Manome, Kazushige Touhara
ABSTRACT
The titan arum, Amorphophallus titanum, is a flowering plant with the largest inflorescence in the world. The flower emits a unique rotting animal-like odor that attracts insects for pollination. To determine the chemical identity of this characteristic odor, we performed gas chromatography-mass spectrometry-olfactometry analysis of volatiles derived from the inflorescence. The main odorant causing the smell during the flower-opening phase was identified as dimethyl trisulfide, a compound with a sulfury odor that has been found to be emitted from some vegetables, microorganisms, and cancerous wounds.
Sigma-Aldrich
Dimethyl trisulfide, ≥98%, FG
Supelco
Dimethyl trisulfide, analytical standard