Merck
  • Biodegradation of leuco derivatives of triphenylmethane dyes by Sphingomonas sp. CM9.
Biodegradation of leuco derivatives of triphenylmethane dyes by Sphingomonas sp. CM9.
Biodegradation (2010-12-29)
Jun Wu, Liguan Li, Hongwei Du, Lijuan Jiang, Qiong Zhang, Zhongbo Wei, Xiaolin Wang, Lin Xiao, Liuyan Yang
ABSTRACT
A leuco derivatives of triphenylmethane dyes degrading bacterium, strain CM9, was isolated from an aquafarm field. Based on morphology, physiologic tests, 16S rDNA sequence, and phylogenetic characteristics, it was identified as Sphingomonas sp. This strain was capable of degrading leucomalachite green (LMG), leucocrystal violet and leucobasic fuchsin completely. The relationship between bacterium growth and LMG degradation suggested that strain CM9 could use LMG as the sole source of carbon. The most LMG degradation activity of CM9 crude extract was observed at pH 7.0 and at 30°C. Many metal ions had little inhibition effect on the degradation activity of the crude extract. CM9 also showed strong decolorization of triphenylmethane dyes to their leuco derivatives. GC/MS analysis detected two novel metabolic products, methylbenzene and 4-aminophenol, during the LMG degradation by CM9.
Sigma-Aldrich
Leucomalachite Green, powder
Supelco
Leucomalachite Green, analytical standard