Merck
  • [Synthesis and investigation on antidiabetic activity of 4-(1-aryl-3-oxo-5-phenylpentylamino) benzenesulfonamide].
[Synthesis and investigation on antidiabetic activity of 4-(1-aryl-3-oxo-5-phenylpentylamino) benzenesulfonamide].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-03-01)
Da-Cheng Yang, Ju-Fang Yan, Jin Xu, Fei Ye, Zu-Wen Zhou, Wei-Yu Zhang, Li Fan, Xin Chen
ABSTRACT
Searching for new antidiabetic lead compound, 4-(1-aryl-3-oxo-5-phenylpentylamino) benzenesulfonamides were designed and synthesized directly by three component one-pot condensation of 4-phenyl-2-butanone and sulfanilamide with some aromatic aldehydes at an yield of 23%-97%. The chemical structures of the twelve new Mannich bases were confirmed by 1H NMR, 13C NMR, FTIR, ESI-MS and HR-MS. The screening results of antidiabetic activity indicated that most of these title compounds possess alpha-glucosidase inhibitory activity, among which compound le is the strongest one. And compound 11 possesses good peroxisome proliferator-activated receptor response element (PPRE) agonist activity. The structure-activity relationship of these new beta-amino ketones containing benzenesulfonamide unit was also discussed preliminarily.
Product Description
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Sulfanilamide, ≥98%