Merck
  • 5-Nitroimidazole-based 1,3,4-thiadiazoles: heterocyclic analogs of metronidazole as anti-Helicobacter pylori agents.
5-Nitroimidazole-based 1,3,4-thiadiazoles: heterocyclic analogs of metronidazole as anti-Helicobacter pylori agents.
Archiv der Pharmazie (2011-03-09)
Mohammad Hassan Moshafi, Maedeh Sorkhi, Saeed Emami, Maryam Nakhjiri, Azadeh Yahya-Meymandi, Amir Soheil Negahbani, Farideh Siavoshi, Maryam Omrani, Eskandar Alipour, Mohsen Vosooghi, Abbas Shafiee, Alireza Foroumadi
ABSTRACT
A series of 5-nitroimidazole-based 1,3,4-thiadiazoles were prepared and tested for antibacterial activity against Helicobacter pylori. The anti-H. pylori activity of target compounds along with the commercially available antimicrobial metronidazole was evaluated by comparing the inhibition-zone diameters determined by the paper disc diffusion bioassay. From our bioassay results against 20 clinical isolates it is evident that piperazinyl, 4-methylpiperazinyl, 3-methylpiperazinyl, and 3,5-dimethylpiperazinyl analogs (6a, 6b, 6e, and 6f, respectively) and pyrrolidine derivative 7 had strong activity at 0.5 µg/disc (average of inhibition zone > 20 mm) while metronidazole had no activity at this dose. Compound 6f containing the 3,5-dimethylpiperazinyl moiety at the 2-position of the 5-(1-methyl-5-nitro-1H-imidazol-2-yl)-1,3,4-thiadiazole skeleton was the most potent compound tested at low concentrations.
Sigma-Aldrich
4-Nitroimidazole, 97%