Merck
  • Paeonia lactiflora Pall inhibits bladder cancer growth involving phosphorylation of Chk2 in vitro and in vivo.
Paeonia lactiflora Pall inhibits bladder cancer growth involving phosphorylation of Chk2 in vitro and in vivo.
Journal of ethnopharmacology (2011-03-15)
Ting-Tsz Ou, Cheng-Hsun Wu, Jeng-Dong Hsu, Charng-Cherng Chyau, Huei-Jane Lee, Chau-Jong Wang
ABSTRACT
Extracts of Paeonia lactiflora Pall (RPA), a traditional Chinese medicines has been shown to treat cancers. The purpose of this study is to evaluate the anticancer effect of RPA in urinary bladder carcinoma in vitro and in vivo. The cell viability was analyzed with DAPI. Flow cytometry and Western blot were used to study the apoptosis and cell cycle related mechanism. A rat model of bladder cancer was induced by N-butyl-N-(4-hydroxybutyl) nitrosamine (OH-BBN). Tumors were analyzed with immunohistochemical analysis. Our data suggested that RPA inhibits growth of bladder cancer via induction of apoptosis and cell cycle arrest. Treatment of TSGH-8301 cells with RPA resulted in G2-M phase arrest that was associated with a marked decline in protein levels of cdc2, cyclin B1, cell division cycle 25B (Cdc25B) and Cdc25C. We also reported that RPA-mediated growth inhibition of TSGH-8301 cells was correlated with activation of checkpoint kinase 2 (Chk2). Herein, we further evaluated urinary bladder cancer using a model of bladder cancer induced by OH-BBN. Analysis of tumors from RPA-treated rats showed significant decrease in the expression of Bcl2, cyclin D1, and PCNA, and increase in the expression of p-Chk2 (Thr-68), Bax, and Cip1/p21. Our data provide the experimental evidence that RPA could modulate apoptosis in models of bladder cancer.
Sigma-Aldrich
N-Butyl-N-(4-hydroxybutyl)nitrosamine, ISOPAC®, ≥90% (GC)
Pricing and availability is not currently available.