Merck
Electrostatic force microscopy of self-assembled peptide structures.
Scanning (2011-04-21)
Casper Hyttel Clausen, Maria Dimaki, Spyros Pantoleon Panagos, Emmanouil Kasotakis, Anna Mitraki, Winnie Edith Svendsen, Jaime Castillo-León
ABSTRACT
In this report electrostatic force microscopy (EFM) is used to study different peptide self-assembled structures such as tubes and particles. It is shown that not only geometrical information can be obtained using EFM, but also information about the composition of different structures. In particular we use EFM to investigate the structures of diphenylalanine peptide tubes, particles, and CSGAITIG peptide particles placed on pre-fabricated SiO(2) surfaces with a backgate. We show that the cavity in the peptide tubes could be due to the presence of water residues. Additionally we show that self-assembled amyloid peptides form spherical solid structures containing the same self-assembled peptide in its interior. In both cases transmission electron microscopy is used to verify these structures. Further, the limitations of the EFM technique are discussed, especially when the observed structures become small compared with the radius of the AFM tip used. Finally, an agreement between the detected signal and the structure of the hollow peptide tubes is demonstrated.
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%