Merck
  • Briefly bound to activate: transient binding of a second catalytic magnesium activates the structure and dynamics of CDK2 kinase for catalysis.
Briefly bound to activate: transient binding of a second catalytic magnesium activates the structure and dynamics of CDK2 kinase for catalysis.
Structure (London, England : 1993) (2011-05-14)
Zhao Qin Bao, Douglas M Jacobsen, Matthew A Young
ABSTRACT
We have determined high-resolution crystal structures of a CDK2/Cyclin A transition state complex bound to ADP, substrate peptide, and MgF(3)(-). Compared to previous structures of active CDK2, the catalytic subunit of the kinase adopts a more closed conformation around the active site and now allows observation of a second Mg(2+) ion in the active site. Coupled with a strong [Mg(2+)] effect on in vitro kinase activity, the structures suggest that the transient binding of the second Mg(2+) ion is necessary to achieve maximum rate enhancement of the chemical reaction, and Mg(2+) concentration could represent an important regulator of CDK2 activity in vivo. Molecular dynamics simulations illustrate how the simultaneous binding of substrate peptide, ATP, and two Mg(2+) ions is able to induce a more rigid and closed organization of the active site that functions to orient the phosphates, stabilize the buildup of negative charge, and shield the subsequently activated γ-phosphate from solvent.
Sigma-Aldrich
Magnesium fluoride, pieces, 3-6 mm, 99.9% trace metals basis (excluding Na)
Sigma-Aldrich
Magnesium fluoride, random crystals, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium fluoride, technical grade