Merck
  • Bile acid-based cage compounds with lipophilic outer shells and inner cavities.
Bile acid-based cage compounds with lipophilic outer shells and inner cavities.
Organic letters (2011-05-20)
Xinyan Bai, Charles Barnes, Robert A Pascal, Xueyi Chen, Jerry Ray Dias
ABSTRACT
The uniquely functionalized steroid-based cyclodimers 4, cis-5, and trans-5 have been synthesized and fully characterized.The cyclodimer 5, with a cis-trans ratio of 3:1, is obtained by coupling the terminal alkenes of two 4-pentenoate groups on a cyclodimer 4 via Grubbs' intramolecular ring-closing metathesis. The crystal structure shows cis-5 to be a cagelike cyclic oligomer bridged by the flexible oct-4-enedioate link.
Sigma-Aldrich
4-Pentenoic acid, 97%
Sigma-Aldrich
4-Pentenoic acid, ≥98%, stabilized, FG