Merck
  • Beta-asarone inhibits synaptic inputs to airway preganglionic parasympathetic motoneurons.
Beta-asarone inhibits synaptic inputs to airway preganglionic parasympathetic motoneurons.
Respiratory physiology & neurobiology (2011-05-31)
Dongying Qiu, Lili Hou, Yonghua Chen, Xujiao Zhou, Wenjun Yuan, Weifang Rong, Lei Zhu, Jijiang Wang
ABSTRACT
Therapeutic application of Asarum, a herbal medicine that has been used for centuries, reportedly causes acute respiratory disturbance. The responsible constituents, the sites of action, and the mechanisms involved in this side effect are unclear. We investigated the effects of β-asarone, a volatile constituent of Asarum, on neurotransmission in the medullary respiratory neuronal network using extracellular recording of the rhythmic hypoglossal activity and voltage clamp recordings of the postsynaptic activity of the airway preganglionic parasympathetic motoneurons (APPMs) in vitro. β-Asarone caused progressive decreases in the duration and area of the hypoglossal bursts in a concentration-dependent manner. The frequency and amplitude of the bursts were initially unaltered or temporarily increased, but were then inhibited progressively after prolonged exposure. As with the inhibition of the hypoglossal bursts, the tonic and the phasic excitatory and inhibitory postsynaptic currents in the APPMs were attenuated. These data suggest that the Asarum-caused acute respiratory disturbance involves β-asarone-induced inhibition of neurotransmission in the medullary respiratory neuronal network.
Sigma-Aldrich
cis-2,4,5-Trimethoxy-1-propenylbenzene, 70%